siRNA Details
| Gene Symbol: Ankrd23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153502.3 | NP_705722.2 | 1 | 33 | ||
| GenBank mRNA | AF492400.1 | - | 1 | 33 | ||
| AK019089.1 | - | 1 | 32 | |||
| BC132370.1 | - | 1 | 43 | 40130647 | ||
| BC138797.1 | - | 1 | 43 | 8861819 | ||
| BC145421.1 | - | 1 | 43 | 9053979 | ||

Ankrd23
Genomic Map