siRNA Details
| Gene Symbol: 1700054A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018858.1 | - | 3 | 429 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700054A03Rik| Gene Symbol: 1700054A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018858.1 | - | 3 | 429 | ||