siRNA Details
| Gene Symbol: Utp23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030132.5 | NP_084408.1 | 3 | 784 | ||
| GenBank mRNA | AK005831.1 | - | 3 | 585 | ||
| AK021306.1 | - | 3 | 734 | |||
| AK042518.1 | - | 3 | 741 | |||
| AK134445.1 | - | 3 | 716 | |||
| BC027540.1 | - | 2 | 545 | |||
| BC046791.1 | - | 3 | 729 | 5717427 | ||
| BC056961.1 | - | 3 | 729 | 5717427 | ||

Utp23
Genomic Map