siRNA Details
| Gene Symbol: Desi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024282.3 | NP_077244.1 | 5 | 899 | ||
| GenBank mRNA | AK020014.1 | - | 5 | 784 | ||
| AK029574.1 | - | 6 | 973 | |||
| AK049579.1 | - | 5 | 668 | |||
| AK146492.1 | - | 5 | 632 | |||
| AK160733.1 | - | 5 | 488 | |||
| BC002200.1 | - | 5 | 568 | 3489351 | ||
| BC046816.1 | - | 5 | 606 | 5702476 | ||

Desi2
Genomic Map