siRNA Details
| Gene Symbol: Cacul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172096.1 | NP_001165567.1 | 1 | 337 | ||
| NM_001172097.1 | NP_001165568.1 | 1 | 337 | |||
| NM_030197.2 | NP_084473.1 | 1 | 337 | |||
| GenBank mRNA | AK019216.1 | - | 1 | 271 | ||
| AK036606.1 | - | 1 | 221 | |||
| AK147382.1 | - | 1 | 340 | |||
| AK169791.1 | - | 1 | 213 | |||
| AK197748.1 | - | 1 | 39 | |||
| BC026363.1 | - | 1 | 307 | 4223313 | ||

Cacul1
Genomic Map