siRNA Details
| Gene Symbol: Gins3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030198.3 | NP_084474.1 | 2 | 427 | ||
| GenBank mRNA | AK019225.1 | - | 2 | 428 | ||
| AK146574.1 | - | 2 | 458 | |||
| AK150602.1 | - | 2 | 459 | |||
| AK151243.1 | - | 2 | 459 | |||
| AK152572.1 | - | 2 | 459 | |||
| AK160606.1 | - | 2 | 428 | |||
| BC039794.1 | - | 2 | 424 | 5121680 | ||
| BC064746.1 | - | 2 | 416 | 30095310 | ||

Gins3
Genomic Map