siRNA Details
| Gene Symbol: Psd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027626.1 | NP_081902.1 | 8 | 8399 | ||
| NM_030263.5 | NP_084539.2 | 13 | 8741 | |||
| NM_177698.4 | NP_808366.2 | 16 | 10505 | |||
| GenBank mRNA | AK032036.1 | - | 1 | 1775 | ||
| BC003498.1 | - | 1 | 253 | |||

Psd3
Genomic Map