siRNA Details
Gene Symbol: Cwc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290740.1 | NP_001277669.1 | 10 | 1364 | ||
NM_030560.5 | NP_085037.2 | 11 | 1422 | |||
NM_172667.2 | NP_766255.1 | 10 | 1361 | |||
GenBank mRNA | AK045589.1 | - | 10 | 1193 | ||
AK077961.1 | - | 11 | 1250 | |||
AK089988.1 | - | 1200 | ||||
AK129404.1 | - | 1178 | ||||
AK132001.1 | - | 10 | 1305 | |||
AK149528.1 | - | 10 | 1188 | |||
AK166487.1 | - | 10 | 1200 | |||
BC003993.1 | - | 10 | 1174 | 3492146 |
