siRNA Details
| Gene Symbol: Fam20c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030565.6 | NP_085042.2 | 9 | 2000 | ||
| GenBank mRNA | AK149780.1 | - | 10 | 2080 | ||
| AK154810.1 | - | 9 | 1996 | |||
| AK220286.1 | - | 9 | 1516 | |||
| AY778962.1 | - | 9 | 1981 | |||
| BC004044.1 | - | 6 | 609 | |||
| BC025814.1 | - | 9 | 1951 | |||
| BC025826.1 | - | 9 | 1981 | |||
| CT010363.1 | - | 5 | 455 | |||

Fam20c
Genomic Map