siRNA Details
| Gene Symbol: Fam20c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030565.6 | NP_085042.2 | 8 | 1921 | ||
| GenBank mRNA | AK149780.1 | - | 9 | 2001 | ||
| AK154810.1 | - | 8 | 1917 | |||
| AK220286.1 | - | 8 | 1437 | |||
| AY778962.1 | - | 8 | 1902 | |||
| BC004044.1 | - | 5 | 530 | |||
| BC025814.1 | - | 8 | 1872 | |||
| BC025826.1 | - | 8 | 1902 | |||
| CT010363.1 | - | 4 | 376 | |||

Fam20c
Genomic Map