siRNA Details
| Gene Symbol: Fam20c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030565.6 | NP_085042.2 | 6, 7 | 1802 | ||
| GenBank mRNA | AK149780.1 | - | 7, 8 | 1882 | ||
| AK154810.1 | - | 6, 7 | 1798 | |||
| AK220286.1 | - | 6, 7 | 1318 | |||
| AY778962.1 | - | 6, 7 | 1783 | |||
| BC004044.1 | - | 3, 4 | 411 | |||
| BC025814.1 | - | 6, 7 | 1753 | |||
| BC025826.1 | - | 6, 7 | 1783 | |||
| CT010363.1 | - | 2, 3 | 257 | |||

Fam20c
Genomic Map