siRNA Details
| Gene Symbol: Hist1h1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030609.3 | NP_085112.1 | 1 | 191 | ||
| GenBank mRNA | BC116820.1 | - | 1 | 166 | 40126139 | |
| BC116850.1 | - | 1 | 164 | 40126169 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h1a| Gene Symbol: Hist1h1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030609.3 | NP_085112.1 | 1 | 191 | ||
| GenBank mRNA | BC116820.1 | - | 1 | 166 | 40126139 | |
| BC116850.1 | - | 1 | 164 | 40126169 | ||