siRNA Details
| Gene Symbol: Trim44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020267.2 | NP_064663.2 | 1 | 772 | ||
| GenBank mRNA | AJ249129.1 | - | 1 | 721 | ||
| AJ278191.1 | - | 1 | 780 | |||
| BC039979.1 | - | 1 | 198 | |||
| BC045602.1 | - | 1 | 187 | |||
| BC098369.1 | - | 1 | 741 | 6830438 | ||

Trim44
Genomic Map