siRNA Details
| Gene Symbol: Vmn1r171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030737.2 | NP_109662.1 | 1 | 1290 | ||
| GenBank mRNA | AF324871.1 | - | 1300 | |||
| BC111911.1 | - | 1 | 1290 | 40046770 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r171| Gene Symbol: Vmn1r171 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030737.2 | NP_109662.1 | 1 | 1290 | ||
| GenBank mRNA | AF324871.1 | - | 1300 | |||
| BC111911.1 | - | 1 | 1290 | 40046770 | ||