siRNA Details
| Gene Symbol: Cacng8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133190.1 | NP_573453.1 | 3 | 489 | ||
| GenBank mRNA | AF361350.1 | - | 3 | 489 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacng8| Gene Symbol: Cacng8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133190.1 | NP_573453.1 | 3 | 489 | ||
| GenBank mRNA | AF361350.1 | - | 3 | 489 | ||