siRNA Details
Gene Symbol: Plekha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031257.3 | NP_112547.1 | 10 | 971 | ||
GenBank mRNA | AF286161.1 | - | 10 | 925 | ||
AF418551.1 | - | 9 | 837 | |||
AK032325.1 | - | 10 | 980 | |||
AK042324.1 | - | 10 | 945 | |||
AK078212.1 | - | 10 | 950 | |||
AK088395.1 | - | 10 | 913 | |||
AK153117.1 | - | 10 | 943 | |||
BC010215.1 | - | 10 | 892 | 3498236 |
