siRNA Details
| Gene Symbol: Plekha2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031257.3 | NP_112547.1 | 12 | 1130 | ||
| GenBank mRNA | AF286161.1 | - | 12 | 1084 | ||
| AF418551.1 | - | 11 | 996 | |||
| AK032325.1 | - | 12 | 1139 | |||
| AK042324.1 | - | 12 | 1104 | |||
| AK078212.1 | - | 12 | 1109 | |||
| AK088395.1 | - | 12 | 1072 | |||
| AK153117.1 | - | 12 | 1102 | |||
| BC010215.1 | - | 12 | 1051 | 3498236 | ||

Plekha2
Genomic Map