siRNA Details
| Gene Symbol: Gsdmc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031378.3 | NP_113555.1 | 9 | 1181 | ||
| GenBank mRNA | AB042406.1 | - | 8 | 1078 | ||
| AK137662.1 | - | 9 | 1182 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gsdmc| Gene Symbol: Gsdmc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031378.3 | NP_113555.1 | 9 | 1181 | ||
| GenBank mRNA | AB042406.1 | - | 8 | 1078 | ||
| AK137662.1 | - | 9 | 1182 | |||