siRNA Details
| Gene Symbol: Smarcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031842.2 | NP_114030.2 | 4 | 468 | ||
| GenBank mRNA | BC026783.1 | - | 4 | 380 | 4481097 | |
| BC059921.1 | - | 4 | 438 | |||
| BC079839.1 | - | 4 | 414 | 6810462 | ||
| CV562378.1 | - | 4 | 468 | |||
| M25773.1 | - | 4 | 424 | |||
| U66620.1 | - | 4 | 424 | |||

Smarcd1
Genomic Map