siRNA Details
| Gene Symbol: Smarcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031842.2 | NP_114030.2 | 7 | 867 | ||
| GenBank mRNA | AK075611.1 | - | 3 | 204 | ||
| AK214257.1 | - | 1 | 13 | |||
| BC026783.1 | - | 7 | 779 | 4481097 | ||
| BC059921.1 | - | 7 | 837 | |||
| BC079839.1 | - | 7 | 813 | 6810462 | ||
| M25773.1 | - | 7 | 823 | |||
| U66620.1 | - | 7 | 823 | |||

Smarcd1
Genomic Map