siRNA Details
| Gene Symbol: Clec2i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289706.1 | NP_001276635.1 | 5 | 467 | ||
| NM_001289707.1 | NP_001276636.1 | 4 | 392 | |||
| NM_001289708.1 | NP_001276637.1 | 3 | 309 | |||
| NM_020257.2 | NP_064653.1 | 4 | 380 | |||
| GenBank mRNA | AF121352.1 | - | 4 | 353 | ||
| AF350411.1 | - | 4 | 357 | |||
| AK041724.1 | - | 3 | 1095 | |||
| AY137344.1 | - | 4 | 391 | |||
| AY137345.1 | - | 4 | 379 | |||
| AY256574.1 | - | 5 | 458 | |||
| AY256575.1 | - | 4 | 381 | |||
| AY256576.1 | - | 3 | 298 | |||
| BC139192.1 | - | 4 | 353 | 8862214 | ||
| BC139193.1 | - | 4 | 353 | 8862215 | ||
| BC145574.1 | - | 4 | 365 | 9054140 | ||
| HQ713440.1 | - | 3 | 287 | |||

Clec2i
Genomic Map