siRNA Details
| Gene Symbol: Pid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003948.2 | NP_001003948.2 | 2, 3 | 454 | ||
| GenBank mRNA | AK035542.1 | - | 2, 3 | 329 | ||
| AK083256.1 | - | 2, 3 | 374 | |||
| AK150971.1 | - | 2, 3 | 392 | |||
| AK151267.1 | - | 2, 3 | 424 | |||
| BC054112.1 | - | 1, 2 | 87 | |||
| BC080290.1 | - | 2, 3 | 400 | 6816865 | ||

Pid1
Genomic Map