siRNA Details
| Gene Symbol: Pid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003948.2 | NP_001003948.2 | 3 | 826 | ||
| GenBank mRNA | AK035542.1 | - | 3 | 701 | ||
| AK083256.1 | - | 3 | 747 | |||
| AK150971.1 | - | 3 | 764 | |||
| AK151267.1 | - | 3 | 796 | |||
| BC054112.1 | - | 2 | 459 | |||
| BC080290.1 | - | 3 | 772 | 6816865 | ||

Pid1
Genomic Map