siRNA Details
| Gene Symbol: Fam102a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153560.4 | NP_705788.1 | 11 | 1881 | ||
| GenBank mRNA | AK049032.1 | - | 11 | 1885 | ||
| AK131145.1 | - | 10 | 3599 | |||
| AK157331.1 | - | 11 | 1511 | |||
| AK157490.1 | - | 12 | 1303 | |||
| AK158435.1 | - | 11 | 1510 | |||
| AK165751.1 | - | 11 | 1516 | |||
| AK170536.1 | - | 11 | 1909 | |||
| AK170847.1 | - | 11 | 1502 | |||
| AK170951.1 | - | 11 | 1508 | |||
| AK172536.1 | - | 11 | 1508 | |||
| BC023470.1 | - | 3 | 214 | |||
| BC023752.1 | - | 11 | 1326 | 5354455 | ||
| BC031157.1 | - | 11 | 1144 | 4976773 | ||

Fam102a
Genomic Map