siRNA Details
| Gene Symbol: Psph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133900.4 | NP_598661.1 | 8 | 1259 | ||
| GenBank mRNA | AA003039.1 | - | 2 | 204 | ||
| AI841354.1 | - | 1 | 292 | |||
| AK088865.1 | - | 8 | 1211 | |||
| AK168048.1 | - | 6 | 867 | |||
| BC002251.1 | - | 6 | 830 | 3493291 | ||

Psph
Genomic Map