siRNA Details
| Gene Symbol: Psph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133900.4 | NP_598661.1 | 6 | 948 | ||
| GenBank mRNA | AK088865.1 | - | 6 | 900 | ||
| AK168048.1 | - | 4 | 556 | |||
| BC002251.1 | - | 4 | 519 | 3493291 | ||
| BQ177662.1 | - | 4 | 514 | |||

Psph
Genomic Map