siRNA Details
| Gene Symbol: Cuedc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172099.1 | NP_001165570.1 | 1 | 464 | ||
| NM_198013.3 | NP_932130.2 | 2 | 662 | |||
| GenBank mRNA | AK049250.1 | - | 2 | 626 | ||
| AK162604.1 | - | 1 | 44 | |||
| AK208246.1 | - | 1 | 55 | |||
| BC024476.1 | - | 2 | 511 | 4976954 | ||
| BC025434.1 | - | 2 | 516 | |||
| BC029708.1 | - | 2 | 595 | 5041450 | ||
| BC131989.1 | - | 1 | 435 | 40130266 | ||
| BC131991.1 | - | 1 | 435 | 40130268 | ||
| BC145206.1 | - | 1 | 435 | 9053757 | ||

Cuedc1
Genomic Map