siRNA Details
| Gene Symbol: Brd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK142152.1 | - | 15 | 1707 | ||
| AK154645.1 | - | 12 | 3947 | |||
| BC031484.1 | - | 15 | 1705 | 2654265 | ||
| BC044769.1 | - | 8 | 1648 | |||
| BC046438.1 | - | 7 | 681 | |||
| BC047372.1 | - | 15 | 1688 | |||
| BC057597.1 | - | 13 | 3273 | |||
| BC089379.1 | - | 15 | 1662 | |||

Brd9
Genomic Map