siRNA Details
Gene Symbol: Brd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142152.1 | - | 13 | 1464 | ||
AK154645.1 | - | 10 | 3704 | |||
BC031484.1 | - | 13 | 1462 | 2654265 | ||
BC044769.1 | - | 6 | 1407 | |||
BC046438.1 | - | 5 | 438 | |||
BC047372.1 | - | 13 | 1445 | |||
BC057597.1 | - | 11 | 3030 | |||
BC089379.1 | - | 13 | 1419 |
