siRNA Details
| Gene Symbol: Dph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047433.2 | NP_001040898.1 | 1 | 237 | ||
| NM_001284346.1 | NP_001271275.1 | 1 | 237 | |||
| NM_172254.4 | NP_758458.1 | 1 | 237 | |||
| GenBank mRNA | AK011448.1 | - | 1 | 155 | ||
| AK017764.1 | - | 1 | 155 | |||
| AK048097.1 | - | 1 | 127 | |||
| BC029910.1 | - | 1 | 128 | 4920779 | ||
| BC039954.1 | - | 1 | 148 | 4193739 | ||
| DN175457.1 | - | 1 | 211 | |||

Dph3
Genomic Map