siRNA Details
| Gene Symbol: Trip10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242389.1 | NP_001229318.1 | 2 | 220 | ||
| NM_001242390.1 | NP_001229319.1 | 2 | 220 | |||
| NM_001242391.1 | NP_001229320.1 | 2 | 220 | |||
| NM_134125.4 | NP_598886.1 | 2 | 220 | |||
| GenBank mRNA | AF502565.1 | - | 2 | 123 | ||
| AK088909.1 | - | 2 | 170 | |||
| AK149902.1 | - | 2 | 192 | |||
| AY081142.1 | - | 2 | 171 | |||
| BC003249.1 | - | 2 | 222 | 3500118 | ||

Trip10
Genomic Map