siRNA Details
| Gene Symbol: Kazald1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178929.4 | NP_849260.2 | 2 | 147 | ||
| GenBank mRNA | AK030678.1 | - | 2 | 146 | ||
| AK039647.1 | - | 1 | 398 | |||
| AK155095.1 | - | 3 | 392 | |||
| BB618180.1 | - | 2 | 146 | |||
| BB871162.1 | - | 2 | 180 | |||
| BC115642.1 | - | 2 | 138 | 40103018 | ||
| BC115643.1 | - | 2 | 135 | |||

Kazald1
Genomic Map