siRNA Details
| Gene Symbol: Ap1s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290378.1 | NP_001277307.1 | 3 | 486 | ||
| NM_001290379.1 | NP_001277308.1 | 3 | 486 | |||
| NM_026887.3 | NP_081163.2 | 3 | 486 | |||
| GenBank mRNA | AK005223.1 | - | 3 | 350 | ||
| AK043776.1 | - | 3 | 307 | |||
| AK045558.1 | - | 3 | 487 | |||
| AK047724.1 | - | 3 | 363 | |||
| AK053941.1 | - | 3 | 359 | |||
| AK167690.1 | - | 3 | 361 | |||
| BC046964.1 | - | 3 | 343 | 6485320 | ||

Ap1s2
Genomic Map