siRNA Details
| Gene Symbol: Ap1s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290378.1 | NP_001277307.1 | 3, 4 | 550 | ||
| NM_001290379.1 | NP_001277308.1 | 3, 4 | 550 | |||
| NM_026887.3 | NP_081163.2 | 3, 4 | 550 | |||
| GenBank mRNA | AK005223.1 | - | 3, 4 | 414 | ||
| AK043776.1 | - | 3, 4 | 371 | |||
| AK045558.1 | - | 3, 4 | 551 | |||
| AK047724.1 | - | 3, 4 | 427 | |||
| AK053941.1 | - | 3, 4 | 423 | |||
| AK167690.1 | - | 3, 4 | 425 | |||
| AK211598.1 | - | 1, 2 | 36 | |||
| BC046964.1 | - | 3, 4 | 407 | 6485320 | ||

Ap1s2
Genomic Map