siRNA Details
| Gene Symbol: Ankrd39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026241.3 | NP_080517.1 | 3 | 403 | ||
| GenBank mRNA | AK018681.1 | - | 3 | 404 | ||
| AK030874.1 | - | 599 | ||||
| AK042896.1 | - | 3 | 404 | |||
| AK081207.1 | - | 3 | 596 | |||
| AK158215.1 | - | 3 | 598 | |||
| AK160231.1 | - | 3 | 419 | |||
| BC117004.1 | - | 3 | 433 | 40126323 | ||
| BC119100.1 | - | 3 | 433 | 8733849 | ||
| BC144893.1 | - | 3 | 433 | |||

Ankrd39
Genomic Map