siRNA Details
| Gene Symbol: Vimp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024439.3 | NP_077759.3 | 3 | 345 | ||
| GenBank mRNA | AF335543.1 | - | 3 | 299 | ||
| AK005204.1 | - | 3 | 328 | |||
| BC011091.1 | - | 3 | 335 | 3983625 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vimp| Gene Symbol: Vimp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024439.3 | NP_077759.3 | 3 | 345 | ||
| GenBank mRNA | AF335543.1 | - | 3 | 299 | ||
| AK005204.1 | - | 3 | 328 | |||
| BC011091.1 | - | 3 | 335 | 3983625 | ||