siRNA Details
| Gene Symbol: Asl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133768.4 | NP_598529.1 | 3 | 267 | ||
| GenBank mRNA | AK010922.1 | - | 3 | 281 | ||
| AK076453.1 | - | 3 | 282 | |||
| AK138423.1 | - | 3 | 264 | |||
| AK140160.1 | - | 3 | 282 | |||
| AK148764.1 | - | 3 | 251 | |||
| AK168739.1 | - | 3 | 262 | |||
| AK185051.1 | - | 1 | 33 | |||
| AK186886.1 | - | 1 | 118 | |||
| BC016670.1 | - | 3 | 267 | 2648753 | ||

Asl
Genomic Map