siRNA Details
| Gene Symbol: Qdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024236.2 | NP_077198.1 | 3, 4 | 404 | ||
| GenBank mRNA | AK011351.1 | - | 3, 4 | 333 | ||
| AK078123.1 | - | 3, 4 | 357 | |||
| AK159107.1 | - | 3, 4 | 348 | |||
| AK161660.1 | - | 3, 4 | 297 | |||
| AK166694.1 | - | 3, 4 | 358 | |||
| AK214053.1 | - | 1, 2 | 14 | |||
| BC002107.1 | - | 3, 4 | 344 | 3487104 | ||

Qdpr
Genomic Map