siRNA Details
| Gene Symbol: Clic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033444.2 | NP_254279.1 | 4 | 523 | ||
| GenBank mRNA | AK088809.1 | - | 4 | 334 | ||
| AK150382.1 | - | 4 | 801 | |||
| AK150611.1 | - | 4 | 512 | |||
| AK153509.1 | - | 4 | 801 | |||
| AK167765.1 | - | 4 | 529 | |||
| AK168262.1 | - | 4 | 343 | |||
| AK186702.1 | - | 2 | 97 | |||
| BC004658.1 | - | 4 | 523 | 3498118 | ||

Clic1
Genomic Map