siRNA Details
| Gene Symbol: Clic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033444.2 | NP_254279.1 | 1 | 218 | ||
| GenBank mRNA | AK088809.1 | - | 1 | 29 | ||
| AK150611.1 | - | 1 | 207 | |||
| AK167765.1 | - | 1 | 224 | |||
| AK168262.1 | - | 1 | 38 | |||
| BC004658.1 | - | 1 | 218 | 3498118 | ||

Clic1
Genomic Map