siRNA Details
| Gene Symbol: Selm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053267.2 | NP_444497.1 | 5 | 665 | ||
| GenBank mRNA | AK018791.1 | - | 5 | 501 | ||
| AK035312.1 | - | 5 | 455 | |||
| AK046034.1 | - | 5 | 457 | |||
| AK079358.1 | - | 5 | 484 | |||
| AK079576.1 | - | 2 | 1275 | |||
| AW049879.1 | - | 1 | 220 | |||
| AY043488.1 | - | 5 | 481 | |||
| BC019742.1 | - | 5 | 357 | |||
| BC147332.1 | - | 5 | 575 | 9056406 | ||
| BC147333.1 | - | 5 | 577 | 9056396 | ||

Selm
Genomic Map