siRNA Details
| Gene Symbol: Selm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053267.2 | NP_444497.1 | 4 | 492 | ||
| GenBank mRNA | AK018791.1 | - | 4 | 328 | ||
| AK035312.1 | - | 4 | 282 | |||
| AK046034.1 | - | 4 | 284 | |||
| AK079576.1 | - | 2 | 1021 | |||
| AW049879.1 | - | 2 | 393 | |||
| AY043488.1 | - | 4 | 308 | |||
| BC019742.1 | - | 4 | 184 | |||
| BC147332.1 | - | 4 | 402 | 9056406 | ||
| BC147333.1 | - | 4 | 404 | 9056396 | ||
| CK787580.1 | - | 4 | 484 | |||

Selm
Genomic Map