siRNA Details
| Gene Symbol: Fgfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164259.1 | NP_001157731.1 | 5 | 1229 | ||
| NM_054071.2 | NP_473412.1 | 6 | 1502 | |||
| GenBank mRNA | AF321300.1 | - | 6 | 1229 | ||
| AF321301.1 | - | 5 | 1190 | |||
| AJ293947.1 | - | 5 | 1135 | |||
| BC058745.1 | - | 5 | 1104 | 6439421 | ||

Fgfrl1
Genomic Map