siRNA Details
| Gene Symbol: Barx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013800.2 | NP_038828.2 | 2 | 644 | ||
| GenBank mRNA | BC012684.1 | - | 2 | 635 | 3980774 | |
| L77900.1 | - | 2 | 442 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Barx2| Gene Symbol: Barx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013800.2 | NP_038828.2 | 2 | 644 | ||
| GenBank mRNA | BC012684.1 | - | 2 | 635 | 3980774 | |
| L77900.1 | - | 2 | 442 | |||