siRNA Details
| Gene Symbol: C1qc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007574.2 | NP_031600.2 | 3 | 654 | ||
| GenBank mRNA | AI385742.1 | - | 1 | 406 | ||
| AK138974.1 | - | 656 | ||||
| BC043945.1 | - | 3 | 634 | 4972540 | ||
| BC054443.1 | - | 3 | 625 | 6475447 | ||
| BC075724.1 | - | 3 | 633 | 3675763 | ||
| X66295.1 | - | 3 | 620 | |||

C1qc
Genomic Map