siRNA Details
| Gene Symbol: Cdk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007668.3 | NP_031694.1 | 10 | 741 | ||
| GenBank mRNA | AK051790.1 | - | 10 | 741 | ||
| AK216613.1 | - | 3 | 133 | |||
| BC052007.1 | - | 10 | 715 | 5706932 | ||
| D29678.1 | - | 10 | 750 | |||

Cdk5
Genomic Map