siRNA Details
Gene Symbol: Cdx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007673.3 | NP_031699.2 | 1 | 299 | ||
GenBank mRNA | AK049566.1 | - | 1 | 300 | ||
AK049574.1 | - | 1 | 171 | |||
BC103516.1 | - | 1 | 177 | 40039012 | ||
BC103517.1 | - | 1 | 180 | 40039015 | ||
BC103519.1 | - | 1 | 180 | 40039019 | ||
BC103520.1 | - | 1 | 180 | 40039017 | ||
S74520.1 | - | 1 | 294 |
