siRNA Details
Gene Symbol: Chrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278041.1 | NP_001264970.1 | 14 | 1752 | ||
NM_009893.2 | NP_034023.1 | 14 | 1686 | |||
GenBank mRNA | AF069501.1 | - | 14 | 1686 | ||
AF096276.1 | - | 14 | 1665 | |||
AK077460.1 | - | 15 | 1847 | |||
AK149445.1 | - | 14 | 1661 | |||
AK165339.1 | - | 14 | 1679 | |||
AK220310.1 | - | 1 | 1216 | |||
BC145017.1 | - | 13 | 1894 | |||
BC145018.1 | - | 15 | 1814 | |||
BC145019.1 | - | 15 | 1851 | |||
BC145696.1 | - | 14 | 1750 | 40130999 |
