siRNA Details
Gene Symbol: Cttn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252572.1 | NP_001239501.1 | 6, 7 | 744 | ||
NM_007803.5 | NP_031829.2 | 6, 7 | 744 | |||
GenBank mRNA | AK084249.1 | - | 6, 7 | 554 | ||
AK146856.1 | - | 6, 7 | 551 | |||
AK148010.1 | - | 6, 7 | 547 | |||
AK165555.1 | - | 6, 7 | 547 | |||
AK168699.1 | - | 6, 7 | 545 | |||
AK168871.1 | - | 6, 7 | 545 | |||
AK168945.1 | - | 6, 7 | 555 | |||
AK177866.1 | - | 1, 2 | 49 | |||
AK184325.1 | - | 1, 2 | 49 | |||
AK189485.1 | - | 1, 2 | 49 | |||
AK206447.1 | - | 1, 2 | 49 | |||
AK216063.1 | - | 1, 2 | 49 | |||
AK216633.1 | - | 1, 2 | 49 | |||
AK219085.1 | - | 1, 2 | 49 | |||
BC011434.1 | - | 6, 7 | 537 | 3981559 | ||
BC117820.1 | - | 5, 6 | 445 | |||
U03184.1 | - | 6, 7 | 533 |
