siRNA Details
Gene Symbol: Cttn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252572.1 | NP_001239501.1 | 6 | 652 | ||
NM_007803.5 | NP_031829.2 | 6 | 652 | |||
GenBank mRNA | AK084249.1 | - | 6 | 462 | ||
AK146856.1 | - | 6 | 459 | |||
AK148010.1 | - | 6 | 455 | |||
AK165555.1 | - | 6 | 455 | |||
AK168699.1 | - | 6 | 453 | |||
AK168871.1 | - | 6 | 453 | |||
AK168945.1 | - | 6 | 463 | |||
AK197295.1 | - | 2 | 57 | |||
BC011434.1 | - | 6 | 445 | 3981559 | ||
BC117820.1 | - | 5 | 353 | |||
U03184.1 | - | 6 | 441 |
